Assay Details
| Gene Symbol: HCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020897.2 | NP_065948.1 | 6 - 7 | 1635 | 57 | |
| NR_073074.1 | 6 - 7 | 1502 | 57 | |||
| XM_011509816.2 | XP_011508118.1 | 6 - 7 | 2153 | 57 | ||
| XM_011509817.2 | XP_011508119.1 | 5 - 6 | 1473 | 57 | ||
| XM_011509818.2 | XP_011508120.1 | 5 - 6 | 865 | 57 | ||
| XM_017001918.1 | XP_016857407.1 | 5 - 6 | 847 | 57 | ||
| GenBank mRNA | AB040968.2 | - | 6 - 7 | 1284 | 57 | |
| AF488550.1 | - | 6 - 7 | 1479 | 57 | ||
| AK299333.1 | - | 6 - 7 | 1475 | 57 | ||
| BC000066.1 | - | 5 - 6 | 863 | 57 | ||
| BC028024.1 | - | 3 - 4 | 500 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001298.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of membrane potential
regulation of postsynaptic membrane potential
cellular response to cAMP
potassium ion transmembrane transport
response to cisplatin
cellular response to dopamine
voltage-gated sodium channel activity
voltage-gated potassium channel activity
cAMP binding
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of membrane potential
regulation of postsynaptic membrane potential
cellular response to cAMP
potassium ion transmembrane transport
response to cisplatin
cellular response to dopamine
voltage-gated sodium channel activity
voltage-gated potassium channel activity
cAMP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01020061.hg.1 | TC01001298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020061.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020061.hg.1 | ENST00000496230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020061.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020060.hg.1 | TC01001298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020060.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020060.hg.1 | ENST00000496230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01020060.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000492035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010121.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | ENST00000496230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173854.hg.1 | TC0100010121.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | ENST00000368358.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | ENST00000467204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | NM_020897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | NR_073074.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100173853.hg.1 | TC0100010121.hg.1 | Clariom D TC |

HCN3
Genomic Map