Assay Details
| Gene Symbol: CNIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073078.1 | 3 - 4 | 481 | 108 | ||
| GenBank mRNA | AY726585.1 | - | 3 - 4 | 226 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB1B, member RAS oncogene family |
RAB1B | -5.2kb | Sense | Chr. 11 - 66268526 - 66277492 on NCBI Build GRCh38 | |
| Downstream |
Yip1 interacting factor homolog A, membrane trafficking protein |
YIF1A |
54TM, FinGER7, YIF1, YIF1P |
1.4kb | Anti-sense | Chr. 11 - 66284580 - 66289167 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
ER to Golgi vesicle-mediated transport
synaptic transmission, glutamatergic
regulation of membrane potential
COPII vesicle coating
localization within membrane
negative regulation of receptor localization to synapse
negative regulation of anterograde synaptic vesicle transport
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
protein binding
ER to Golgi vesicle-mediated transport
synaptic transmission, glutamatergic
regulation of membrane potential
COPII vesicle coating
localization within membrane
negative regulation of receptor localization to synapse
negative regulation of anterograde synaptic vesicle transport
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | ENST00000534826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008069.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000311445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000528063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000528852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000530519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | ENST00000531936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | NM_182553.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | NR_073078.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | NR_073079.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158905.hg.1 | TC1100008069.hg.1 | Clariom D TC |

CNIH2
Genomic Map