Assay Details
| Gene Symbol: SERPINB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073111.1 | 3 - 4 | 543 | 85 | ||
| GenBank mRNA | AK298044.1 | - | 3 - 4 | 354 | 85 | |
| AK309765.1 | - | 3 - 4 | 543 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Werner helicase interacting protein 1 | WRNIP1 |
WHIP, bA420G6.2 |
-52.8kb | Anti-sense | Chr. 6 - 2765341 - 2785745 on NCBI Build GRCh38 |
| Downstream | microRNA 4645 | MIR4645 | mir-4645 | 14.6kb | Sense | Chr. 6 - 2854031 - 2854107 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK298044.1 transcripts.
Set Membership: |
|

SERPINB1
Genomic Map