Assay Details
| Gene Symbol: NEDD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142393.1 | NP_001135865.1 | 6 - 7 | 2259 | 70 | |
| NM_001271033.1 | NP_001257962.1 | 4 - 5 | 1617 | 70 | ||
| NM_006403.3 | NP_006394.1 | 5 - 6 | 2064 | 70 | ||
| NR_073131.1 | 8 - 9 | 2504 | 70 | |||
| GenBank mRNA | AK292682.1 | - | 6 - 7 | 2259 | 70 | |
| BC040207.1 | - | 5 - 6 | 2050 | 4820745 | 70 | |
| BX648041.1 | - | 8 - 9 | 2468 | 70 | ||
| L43821.1 | - | 5 - 6 | 2060 | 70 | ||
| U64317.1 | - | 5 - 6 | 2035 | 70 | ||
More Information
Set Membership: |
|

NEDD9
Genomic Map