Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010589.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010589.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010589.hg.1 |
ENST00000627649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010589.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010589.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010589.hg.1 |
NR_031623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019011.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019011.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019011.hg.1 |
NR_031623.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019011.hg.1 |
TC12001453.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
ENST00000627649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
NR_031623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
ENST00000627649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
NR_031623.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180142.hg.1 |
TC1200010589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010589.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010589.hg.1 |
ENST00000408564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010589.hg.1 |
ENST00000627649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010589.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010589.hg.1 |
NR_002968.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010589.hg.1 |
NR_031623.1 |
Refseq |