Assay Details
| Gene Symbol: RPL23AP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024531.1 | 2 - 3 | 221 | 96 | ||
| GenBank mRNA | AW340513.1 | - | 1 - 2 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L23a pseudogene 82 | RPL23AP82 | RPL23A_43_1761 | Anti-sense | Chr. 22 - 50757086 - 50799637 on NCBI Build GRCh38 | |
| Downstream |
RAB, member of RAS oncogene family-like 2A |
RABL2A | 1.5kb | Anti-sense | Chr. 2 - 113627218 - 113643398 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002227.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02036155.hg.1 | TC02002227.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036155.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036155.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036155.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036155.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036155.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | TC02002227.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | ENST00000416673.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036150.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | ENST00000449021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036148.hg.1 | TC02002227.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036148.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036148.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202279.hg.1 | TC0200013938.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | ENST00000449021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013938.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202277.hg.1 | TC0200013938.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200202265.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202265.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202265.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202265.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202265.hg.1 | TC0200013938.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | ENST00000391616.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | ENST00000416673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_000029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024528.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024529.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202271.hg.1 | TC0200013938.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200202266.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202266.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202266.hg.1 | NR_024530.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202266.hg.1 | NR_024531.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200202266.hg.1 | TC0200013938.hg.1 | Clariom D TC |

RPL23AP7
Genomic Map