Assay Details
Gene Symbol: TNRC6C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040071.1 | 1 | 2523 | 126 | ||
GenBank mRNA | AK094898.1 | - | 1 | 1995 | 126 | |
AK127699.1 | - | 1 | 564 | 126 | ||
AY129024.1 | - | 1 | 1059 | 126 | ||
BC041417.2 | - | 1 | 2524 | 126 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | trinucleotide repeat containing 6C | TNRC6C | -216.0 | Anti-sense | Chr. 17 - 77974984 - 78108835 on NCBI Build GRCh38 | |
Downstream | transmembrane channel like 6 | TMC6 |
EV1, EVER1, EVIN1, LAK-4P |
1.1kb | Sense | Chr. 17 - 78108775 - 78132427 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001912.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P857624 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P19000 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P857624 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P19000 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 23759 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 30105 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57573 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20661 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P19000 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P857624 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03295059 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P857624 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P19000 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03295059 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 230526_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 239062_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 242364_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR17025902.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17025902.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025902.hg.1 | TC17001912.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17025900.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17025900.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025900.hg.1 | TC17001912.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012480.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025903.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025903.hg.1 | TC17001912.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208187.hg.1 | TC1700012480.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208185.hg.1 | TC1700012480.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000374976.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | ENST00000592939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012480.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208188.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208188.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208188.hg.1 | ENST00000589217.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700208188.hg.1 | NR_040071.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700208188.hg.1 | TC1700012480.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 230526_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 242364_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 239062_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 228591_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 239062_at | - | - |
Human Genome U133 Plus 2.0 Array | 230526_at | - | - |
Human Genome U133 Plus 2.0 Array | 228591_at | - | - |
Human Genome U133 Plus 2.0 Array | 242364_x_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239062_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230526_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228591_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242364_x_at8 | - | - |
Human Genome U95 Set | 54483_PM_at | - | - |
Human Genome U95 Set | 54483_at | - | - |
Human Genome U95 Set | 57763_PM_at | - | - |
Human Genome U95 Set | 57763_at | - | - |
Human Genome U95 Set | 91502_PM_at | - | - |
Human Genome U95 Set | 91502_at | - | - |
Human Genome U95 Set | 89316_PM_at | - | - |
Human Genome U95 Set | 89316_at | - | - |
Human Genome U133 Set | 239062_PM_at | - | - |
Human Genome U133 Set | 230526_PM_at | - | - |
Human Genome U133 Set | 239062_at | - | - |
Human Genome U133 Set | 230526_at | - | - |
Human Genome U133 Set | 228591_at | - | - |
Human Genome U133 Set | 228591_PM_at | - | - |
Human Genome U133 Set | 242364_x_at | - | - |
Human Genome U133 Set | 242364_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008836 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008836 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 232965 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3296943 | AK123771.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3296943 | AK127699.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3216365 | AY129024.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3216365 | AK123771.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3216365 | AK127699.1 | human_transcriptome |
