Assay Details
| Gene Symbol: SH3GL1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033420.1 | 1 | 48 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 37B pseudogene 1 | LRRC37BP1 | LRRC37B2 | Sense | Chr. 17 - 30631755 - 30637477 on NCBI Build GRCh38 | |
| Downstream | SUZ12 polycomb repressive complex 2 subunit pseudogene 1 | SUZ12P1 | SUZ12P | 82.8kb | Sense | Chr. 17 - 30709608 - 30770050 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000353.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000590010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000579186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000579186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000582640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000582640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000582640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000583601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000583601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | ENST00000583601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007461.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007461.hg.1 | ENST00000582245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007461.hg.1 | NR_033420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000435469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000435469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000435469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000590734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000590734.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000613178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000620266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000620266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000620266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000622111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000585915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000586022.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000590010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011551.hg.1 | ENST00000590019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000435469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000435469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000435469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000590734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000590734.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000613178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000620266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000620266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000620266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000622111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | ENST00000622750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008721.hg.1 | NR_027418.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000579186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000579186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000582640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000582640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000582640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000583601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000583601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | ENST00000583601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | NR_033412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007500.hg.1 | NR_033412.1 | Refseq |

SH3GL1P2
Genomic Map