Assay Details
Gene Symbol: TFPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271003.1 | NP_001257932.1 | 1 - 2 | 406 | 110 | |
GenBank mRNA | AK092499.1 | - | 1 - 2 | 168 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
