Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261837.1 | NP_001248766.1 | 4 - 5 | 992 | 73 | |
| XM_017016928.1 | XP_016872417.1 | 3 - 4 | 952 | 73 | ||
| XM_017016929.1 | XP_016872418.1 | 3 - 4 | 858 | 73 | ||
| XM_017016930.1 | XP_016872419.1 | 3 - 4 | 829 | 73 | ||
| GenBank mRNA | AK095486.1 | - | 4 - 5 | 952 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000485788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | ENST00000535784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006880.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147131.hg.1 | TC1000006880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | ENST00000298942.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | ENST00000378000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | ENST00000423462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001001484.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001261836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001261837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_001261838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | NM_030664.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147129.hg.1 | TC1000006880.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 183785 | - | - |

PTER
Genomic Map