Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007470.hg.1 |
ENST00000258648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007470.hg.1 |
ENST00000378586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007470.hg.1 |
ENST00000378594.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007470.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007470.hg.1 |
TC13000653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007471.hg.1 |
ENST00000258648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007471.hg.1 |
ENST00000378586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007471.hg.1 |
ENST00000378594.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007471.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007471.hg.1 |
TC13000653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000495013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000495013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
ENST00000495013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007472.hg.1 |
ENST00000258648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007472.hg.1 |
ENST00000378586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007472.hg.1 |
ENST00000378594.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007472.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007472.hg.1 |
TC13000653.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159950.hg.1 |
TC1300008879.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000258648.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000378586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000417167.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000495013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000495013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
ENST00000495013.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_001270629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008879.hg.1 |
NM_014166.3 |
Refseq |