Assay Details
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270735.1 | NP_001257664.1 | 3 - 4 | 584 | 72 | |
GenBank mRNA | DA169152.1 | - | 3 - 4 | 433 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
