Assay Details
| Gene Symbol: CAMKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024766.4 | NP_079042.1 | 6 - 7 | 654 | 104 | |
| XM_011533111.2 | XP_011531413.1 | 6 - 7 | 620 | 104 | ||
| XM_011533113.2 | XP_011531415.1 | 4 - 5 | 330 | 104 | ||
| XM_017004982.1 | XP_016860471.1 | 4 - 5 | 2757 | 104 | ||
| XM_017004983.1 | XP_016860472.1 | 5 - 6 | 869 | 104 | ||
| GenBank mRNA | AK027104.1 | - | 6 - 7 | 594 | 104 | |
| BC053733.1 | - | 6 - 7 | 581 | 5199179 | 104 | |
More Information
Set Membership: |
|

CAMKMT
Genomic Map