Assay Details
Gene Symbol: MED27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253882.1 | NP_001240811.1 | 2 - 3 | 395 | 106 | |
GenBank mRNA | BU069864.1 | - | 1 - 2 | 106 | ||
CD388680.1 | - | 2 - 3 | 402 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001671.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | TC09001671.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | ENST00000292035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | ENST00000357028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | ENST00000372184.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021288.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021286.hg.1 | TC09001671.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09021286.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021286.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | TC09001671.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | ENST00000292035.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | ENST00000357028.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | ENST00000372184.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09021287.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180569.hg.1 | TC0900011793.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900180562.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180562.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180562.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180562.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180562.hg.1 | TC0900011793.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180568.hg.1 | TC0900011793.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180564.hg.1 | TC0900011793.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000292035.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000357028.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000444872.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011793.hg.1 | NM_004269.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180561.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180561.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180561.hg.1 | ENST00000474263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180561.hg.1 | NM_001253882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180561.hg.1 | TC0900011793.hg.1 | Clariom D TC |
