Assay Details
| Gene Symbol: MLL-MLL fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF024540.1 | - | 113 | 136 | ||
| AY187942.1 | - | 150 | 136 | |||
| AY187944.1 | - | 279 | 268 | |||
More Information
Set Membership: |
|

MLL-MLL fusion
Genomic Map