Assay Details
Gene Symbol: LOC101929709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125822.1 | 4 - 5 | 1268 | 93 | ||
GenBank mRNA | BX384738.2 | - | 1 - 2 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | matrix metallopeptidase 16 | MMP16 |
C8orf57, MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP |
-1.4mb | Sense | Chr. 8 - 88037232 - 88327488 on NCBI Build GRCh38 |
Downstream | receptor interacting serine/threonine kinase 2 | RIPK2 |
CARD3, CARDIAK, CCK, GIG30, RICK, RIP2 |
35.6kb | Anti-sense | Chr. 8 - 89757747 - 89791064 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519655.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000519854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000520306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524166.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000524190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012438.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000521996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | ENST00000523995.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187634.hg.1 | TC0800012438.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | ENST00000504145.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | ENST00000621883.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | NR_125822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187626.hg.1 | TC0800012438.hg.1 | Clariom D TC |
