Assay Details
Gene Symbol: LOC101928304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125855.1 | 1 - 2 | 107 | 63 | ||
NR_125856.1 | 1 - 2 | 107 | 63 | |||
GenBank mRNA | AV705921.2 | - | 1 - 2 | 140 | 63 | |
DA256978.1 | - | 1 - 2 | 94 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1010 | LINC01010 | -26.8kb | Anti-sense | Chr. 6 - 134437716 - 134504020 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928277 | LOC101928277 | 133.9kb | Sense | Chr. 6 - 134673910 - 134879364 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202647.hg.1 | TC0600014342.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202654.hg.1 | TC0600014342.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202655.hg.1 | TC0600014342.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202642.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202642.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202642.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202642.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202642.hg.1 | TC0600014342.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600202649.hg.1 | TC0600014342.hg.1 | Clariom D TC |
