Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202636.hg.1 |
TC0600014342.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000606039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014342.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000418837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000423028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
ENST00000607531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
NR_125855.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
NR_125856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202638.hg.1 |
TC0600014342.hg.1 |
Clariom D TC |