Assay Details
Gene Symbol: NAMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ673931.1 | - | 1 - 2 | 72 | 64 | |
DQ673932.1 | - | 1 - 2 | 71 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Sec61 translocon beta subunit | SEC61B | -135.1kb | Anti-sense | Chr. 9 - 99222288 - 99230619 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928438 | LOC101928438 | 219.3kb | Sense | Chr. 9 - 99585786 - 99819889 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
