Assay Details
| Gene Symbol: DLGAP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX647607.1 | - | 1 | 1625 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
gastric adenocarcinoma associated, positive CD44 regulator, long intergenic non-coding RNA |
GAPLINC | LINC01540 | -116.7kb | Anti-sense | Chr. 18 - 3466250 - 3478978 on NCBI Build GRCh38 |
| Downstream | DLGAP1 antisense RNA 2 | DLGAP1-AS2 | 7.2kb | Anti-sense | Chr. 18 - 3603737 - 3610089 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DLGAP1-AS1
Genomic Map