Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
TC2200008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160378.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
TC2200008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160381.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
TC2200008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160377.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
TC2200008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160379.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
TC2200008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160380.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |