Assay Details
Gene Symbol: HORMAD2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110541.1 | 2 - 3 | 486 | 104 | ||
GenBank mRNA | BG618381.1 | - | 2 - 3 | 187 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myotubularin related protein 3 | MTMR3 |
FYVE-DSP1, ZFYVE10 |
-9.3kb | Anti-sense | Chr. 22 - 29883165 - 30030868 on NCBI Build GRCh38 |
Downstream | HORMA domain containing 2 | HORMAD2 | CT46.2 | 29.5kb | Anti-sense | Chr. 22 - 30080069 - 30207425 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | TC2200008418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160367.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | TC2200008418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160368.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | TC2200008418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160374.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | TC2200008418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200160371.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000429350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000432568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000453743.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | ENST00000624945.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008418.hg.1 | NR_110541.1 | Refseq |
