Assay Details
Gene Symbol: LOC101929908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK310325.1 | - | 2 - 3 | 176 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 93 | CCDC93 | Anti-sense | Chr. 2 - 117915478 - 118014163 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
