Assay Details
Gene Symbol: TMPRSS2-ERG fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_TMPRSS2-ERG_T3E4_COSF30.0 | - | 54 | 75 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMPRSS2-ERG fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_TMPRSS2-ERG_T3E4_COSF30.0 | - | 54 | 75 |