Assay Details
| Gene Symbol: KIAA1549-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KIAA1549-BRAF_K17B10_COSF509.0 | - | 47 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1549-BRAF fusion| Gene Symbol: KIAA1549-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KIAA1549-BRAF_K17B10_COSF509.0 | - | 47 | 97 | ||