Assay Details
Gene Symbol: FGFR2-BICC1 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB821310.1 | - | 2019 | 88 | ||
OM_FGFR2-BICC1_F17B3.0 | - | 52 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FGFR2-BICC1 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB821310.1 | - | 2019 | 88 | ||
OM_FGFR2-BICC1_F17B3.0 | - | 52 | 88 |