Assay Details
| Gene Symbol: B4GALT1-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_B4GALT1-RAF1_B1R8.0 | - | 71 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4GALT1-RAF1 fusion| Gene Symbol: B4GALT1-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_B4GALT1-RAF1_B1R8.0 | - | 71 | 82 | ||