Assay Details
| Gene Symbol: CEP85L-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CEP85L-ROS1_C8R36.0 | - | 48 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEP85L-ROS1 fusion| Gene Symbol: CEP85L-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CEP85L-ROS1_C8R36.0 | - | 48 | 110 | ||
Set Membership: |
|