Assay Details
| Gene Symbol: IRF2BP2-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_IRF2BP2-NTRK1_I1N10.0 | - | 76 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRF2BP2-NTRK1 fusion| Gene Symbol: IRF2BP2-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_IRF2BP2-NTRK1_I1N10.0 | - | 76 | 109 | ||
Set Membership: |
|