Assay Details
Gene Symbol: KRT78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300814.1 | NP_001287743.1 | 4 - 5 | 436 | 101 | |
NM_173352.3 | NP_775487.2 | 4 - 5 | 818 | 101 | ||
GenBank mRNA | AK096419.1 | - | 4 - 5 | 818 | 101 | |
AK290901.1 | - | 4 - 5 | 818 | 101 | ||
AK298636.1 | - | 5 - 6 | 744 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001538.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12020208.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020208.hg.1 | ENST00000359499.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020208.hg.1 | ENST00000539860.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020208.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020208.hg.1 | TC12001538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182254.hg.1 | TC1200010778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
