Assay Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300848.1 | NP_001287777.1 | 2 - 3 | 432 | 73 | |
| NM_015950.4 | NP_057034.2 | 2 - 3 | 432 | 73 | ||
| XM_005249161.4 | XP_005249218.1 | 2 - 3 | 253 | 73 | ||
| GenBank mRNA | AF132956.1 | - | 2 - 3 | 306 | 73 | |
| AK301041.1 | - | 2 - 3 | 289 | 73 | ||
| AK314948.1 | - | 2 - 3 | 290 | 73 | ||
| BC013685.1 | - | 2 - 3 | 277 | 3862065 | 73 | |
| BC020212.1 | - | 2 - 3 | 277 | 4641018 | 73 | |
| BM563754.1 | - | 398 | 73 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001743.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06020832.hg.1 | TC06001743.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020832.hg.1 | ENST00000230413.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020832.hg.1 | ENST00000388752.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020832.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020833.hg.1 | TC06001743.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020833.hg.1 | ENST00000230413.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020833.hg.1 | ENST00000388752.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020833.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181411.hg.1 | TC0600011859.hg.1 | Clariom D TC |

MRPL2
Genomic Map