Assay Details
Gene Symbol: C5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300894.1 | NP_001287823.1 | 2 - 3 | 673 | 86 | |
GenBank mRNA | AJ437658.2 | - | 2 - 3 | 545 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194138.hg.1 | TC0500013229.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000435259.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000507390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | ENST00000508791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013229.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194145.hg.1 | TC0500013229.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194147.hg.1 | TC0500013229.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000507390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000338051.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000394976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000504727.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | ENST00000507390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | NM_001135586.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | NM_001300894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | NM_152409.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500194142.hg.1 | TC0500013229.hg.1 | Clariom D TC |
