Assay Details
| Gene Symbol: MRPS34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300900.1 | NP_001287829.1 | 2 - 3 | 378 | 130 | |
| GenBank mRNA | EF036486.1 | - | 2 - 3 | 358 | 130 | |
More Information
Set Membership: |
|

MRPS34
Genomic Map