Assay Details
| Gene Symbol: NPSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300933.1 | NP_001287862.1 | 6 - 7 | 900 | 109 | |
| NM_001300934.1 | NP_001287863.1 | 4 - 5 | 735 | 109 | ||
| NM_001300935.1 | NP_001287864.1 | 6 - 7 | 933 | 109 | ||
| NM_207172.1 | NP_997055.1 | 6 - 7 | 887 | 109 | ||
| NM_207173.1 | NP_997056.1 | 6 - 7 | 887 | 109 | ||
| GenBank mRNA | AY310326.1 | - | 6 - 7 | 887 | 109 | |
| AY310327.1 | - | 6 - 7 | 887 | 109 | ||
| AY310328.1 | - | 6 - 7 | 854 | 109 | ||
| AY310330.1 | - | 5 - 6 | 783 | 109 | ||
| AY310331.1 | - | 5 - 6 | 793 | 109 | ||
| AY310332.1 | - | 4 - 5 | 689 | 109 | ||
| AY956446.1 | - | 6 - 7 | 792 | 109 | ||
| AY956447.1 | - | 6 - 7 | 763 | 109 | ||
| AY956448.1 | - | 6 - 7 | 763 | 109 | ||
| BC132693.1 | - | 6 - 7 | 804 | 40146715 | 109 | |
| BC132695.1 | - | 6 - 7 | 804 | 40146717 | 109 | |
| BC144380.1 | - | 4 - 5 | 606 | 9052901 | 109 | |
More Information
Set Membership: |
|

NPSR1
Genomic Map