Assay Details
| Gene Symbol: CCDC28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301011.1 | NP_001287940.1 | 3 - 4 | 673 | 129 | |
| NM_024296.4 | NP_077272.2 | 3 - 4 | 673 | 129 | ||
| XM_011542113.2 | XP_011540415.1 | 3 - 4 | 1432 | 129 | ||
| XM_017002306.1 | XP_016857795.1 | 3 - 4 | 673 | 129 | ||
| XM_017002307.1 | XP_016857796.1 | 3 - 4 | 1427 | 129 | ||
| GenBank mRNA | AK225113.1 | - | 3 - 4 | 443 | 129 | |
| AK291904.1 | - | 3 - 4 | 683 | 129 | ||
| BC022848.2 | - | 3 - 4 | 407 | 3946778 | 129 | |
More Information
Set Membership: |
|

CCDC28B
Genomic Map