Assay Details
| Gene Symbol: C11orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082969.1 | NP_001076438.1 | 3 - 4 | 288 | 89 | |
| NM_001082970.1 | NP_001076439.1 | 3 - 4 | 846 | 89 | ||
| NM_001301017.1 | NP_001287946.1 | 3 - 4 | 288 | 89 | ||
| NM_001301019.1 | NP_001287948.1 | 3 - 4 | 329 | 89 | ||
| NM_001301021.1 | NP_001287950.1 | 3 - 4 | 329 | 89 | ||
| NM_018195.3 | NP_060665.3 | 3 - 4 | 846 | 89 | ||
| NR_103469.1 | 3 - 4 | 887 | 89 | |||
| GenBank mRNA | AB096251.1 | - | 3 - 4 | 192 | 89 | |
| AK094203.1 | - | 3 - 4 | 222 | 89 | ||
| AK125340.1 | - | 3 - 4 | 802 | 89 | ||
| BC005403.1 | - | 1 - 2 | 85 | 89 | ||
| BC030546.1 | - | 1 - 2 | 90 | 89 | ||
| BC048133.1 | - | 3 - 4 | 774 | 5295844 | 89 | |
| BC072455.1 | - | 3 - 4 | 213 | 6185048 | 89 | |
| BG115155.1 | - | 3 - 4 | 336 | 89 | ||
| BX538107.1 | - | 3 - 4 | 845 | 89 | ||
| CD244552.1 | - | 3 - 4 | 247 | 89 | ||
| DA813095.1 | - | 3 - 4 | 274 | 89 | ||
| EL736728.1 | - | 1 - 2 | 78 | 89 | ||
More Information
Set Membership: |
|

C11orf57
Genomic Map