Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
TC07001505.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
ENST00000222800.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
ENST00000322862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
ENST00000395147.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
ENST00000437775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
ENST00000458339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
NM_001145364.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
NM_148912.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
NM_148913.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022670.hg.1 |
NR_026912.2 |
Refseq |