Assay Details
| Gene Symbol: POLR2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301130.1 | NP_001288059.1 | 2 - 3 | 220 | 116 | |
| NM_001301131.1 | NP_001288060.1 | 2 - 3 | 220 | 116 | ||
| NM_021974.4 | NP_068809.1 | 2 - 3 | 220 | 116 | ||
| XM_017028832.1 | XP_016884321.1 | 2 - 3 | 186 | 116 | ||
| GenBank mRNA | BC003582.1 | - | 2 - 3 | 198 | 3546712 | 116 |
| BP195499.1 | - | 2 - 3 | 186 | 116 | ||
| CR456546.1 | - | 2 - 3 | 110 | 116 | ||
| CR457021.1 | - | 2 - 3 | 95 | 116 | ||
| DA372532.1 | - | 2 - 3 | 220 | 116 | ||
| DN989996.1 | - | 2 - 3 | 195 | 116 | ||
More Information
Set Membership: |
|

POLR2F
Genomic Map