Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301203.1 | NP_001288132.1 | 2 - 3 | 610 | 98 | |
| GenBank mRNA | AK304148.1 | - | 2 - 3 | 516 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | ENST00000561560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157568.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157568.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157568.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157568.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157568.hg.1 | TC1500008003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157567.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157567.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157567.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157567.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157567.hg.1 | TC1500008003.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000299705.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000424155.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000536821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000543455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000558562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | ENST00000561560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_001301203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NM_007364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008003.hg.1 | NR_125394.1 | Refseq |

TMED3
Genomic Map