| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
ENST00000278025.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
ENST00000368552.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
ENST00000447176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
ENST00000465384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023627.hg.1 |
TC10001789.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012398.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000278025.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000368551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000368552.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000447176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000465384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
ENST00000478895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_001098483.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_001301827.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_001301828.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
NM_198472.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000184634.hg.1 |
TC1000012398.hg.1 |
Clariom D TC |