Assay Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015950.4 | NP_057034.2 | 6 - 7 | 868 | 73 | |
| XM_005249161.4 | XP_005249218.1 | 6 - 7 | 689 | 73 | ||
| GenBank mRNA | AF132956.1 | - | 6 - 7 | 742 | 73 | |
| AK314948.1 | - | 6 - 7 | 726 | 73 | ||
| BC013685.1 | - | 6 - 7 | 713 | 3862065 | 73 | |
| BC020212.1 | - | 6 - 7 | 713 | 4641018 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001743.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06020818.hg.1 | TC06001743.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020818.hg.1 | ENST00000388752.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020818.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181398.hg.1 | TC0600011859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181400.hg.1 | TC0600011859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000468957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000470667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000480286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000487429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | ENST00000491898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011859.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181395.hg.1 | TC0600011859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000230413.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000388752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000485654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | ENST00000489623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | NM_001300848.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | NM_015950.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181399.hg.1 | TC0600011859.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 215824 | - | - |

MRPL2
Genomic Map