Assay Details
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127889.4 | NP_001121361.1 | 3 - 4 | 347 | 106 | |
NM_016630.6 | NP_057714.1 | 3 - 4 | 528 | 106 | ||
XM_005254437.4 | XP_005254494.1 | 4 - 5 | 561 | 106 | ||
XM_017022297.1 | XP_016877786.1 | 3 - 4 | 896 | 106 | ||
XM_017022298.1 | XP_016877787.1 | 4 - 5 | 986 | 106 | ||
GenBank mRNA | AF208861.1 | - | 2 - 3 | 243 | 106 | |
AF212231.1 | - | 3 - 4 | 315 | 106 | ||
AK123725.1 | - | 1 - 2 | 760 | 106 | ||
AK172849.1 | - | 3 - 4 | 360 | 106 | ||
AK225527.1 | - | 3 - 4 | 360 | 106 | ||
AK312943.1 | - | 3 - 4 | 362 | 106 | ||
BC000244.1 | - | 3 - 4 | 398 | 3357388 | 106 | |
BI552731.1 | - | 3 - 4 | 498 | 106 | ||
BP268459.1 | - | 3 - 4 | 521 | 106 | ||
BP268875.1 | - | 3 - 4 | 106 | |||
DA680695.1 | - | 3 - 4 | 346 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
