Assay Details
| Gene Symbol: PCDH11X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168362.1 | NP_001161834.1 | 4 - 5 | 3299 | 82 | |
| NM_032968.4 | NP_116750.1 | 6 - 7 | 3410 | 82 | ||
| NM_032969.4 | NP_116751.1 | 5 - 6 | 3380 | 82 | ||
| XM_011530910.1 | XP_011529212.1 | 9 - 10 | 3885 | 82 | ||
| XM_011530911.2 | XP_011529213.1 | 11 - 12 | 4173 | 82 | ||
| XM_011530914.2 | XP_011529216.1 | 7 - 8 | 3785 | 82 | ||
| XM_017029417.1 | XP_016884906.1 | 10 - 11 | 4119 | 82 | ||
| XM_017029418.1 | XP_016884907.1 | 9 - 10 | 4089 | 82 | ||
| XM_017029420.1 | XP_016884909.1 | 8 - 9 | 4008 | 82 | ||
| GenBank mRNA | AB037747.1 | - | 5 - 6 | 598 | 82 | |
| AF332218.1 | - | 6 - 7 | 4211 | 82 | ||
| AF332219.1 | - | 5 - 6 | 4181 | 82 | ||
More Information
Set Membership: |
|

PCDH11X
Genomic Map