Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000366927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000366928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000460522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000463964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000469590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000474379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000475724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000477938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000478794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000481007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000483635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300772.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_138794.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000366927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000366928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000460522.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000463964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000469590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000474379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000475724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000477938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000478794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000481007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
ENST00000483635.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300769.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_001300772.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
NM_138794.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240376.hg.1 |
TC0100018344.hg.1 |
Clariom D TC |