Assay Details
Gene Symbol: LYPLAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300769.1 | NP_001287698.1 | 3 - 4 | 384 | 140 | |
NM_001300770.1 | NP_001287699.1 | 4 - 5 | 489 | 140 | ||
NM_001300771.1 | NP_001287700.1 | 4 - 5 | 520 | 140 | ||
NM_001300772.1 | NP_001287701.1 | 4 - 5 | 438 | 140 | ||
NM_138794.4 | NP_620149.2 | 4 - 5 | 537 | 140 | ||
XM_006711162.3 | XP_006711225.1 | 5 - 6 | 516 | 140 | ||
XM_011509182.2 | XP_011507484.1 | 5 - 6 | 548 | 140 | ||
XM_011509183.2 | XP_011507485.1 | 5 - 6 | 849 | 140 | ||
XM_017000271.1 | XP_016855760.1 | 5 - 6 | 548 | 140 | ||
XM_017000272.1 | XP_016855761.1 | 4 - 5 | 753 | 140 | ||
GenBank mRNA | AK123628.1 | - | 3 - 4 | 329 | 140 | |
AK129913.1 | - | 4 - 5 | 341 | 140 | ||
AK291542.1 | - | 4 - 5 | 491 | 140 | ||
AK309689.1 | - | 3 - 4 | 346 | 140 | ||
AY341430.1 | - | 4 - 5 | 429 | 140 | ||
BC016711.1 | - | 4 - 5 | 491 | 4133970 | 140 | |
CB851909.1 | - | 4 - 5 | 527 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
