Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000433347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
ENST00000433347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165835.hg.1 |
TC0900009604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165837.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165837.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165837.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165837.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165837.hg.1 |
TC0900009604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000380685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000433347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
ENST00000433347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NM_001271829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009604.hg.1 |
NR_073471.1 |
Refseq |