Assay Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286703.1 | NP_001273632.1 | 5 - 6 | 330 | 108 | |
| NM_001286704.1 | NP_001273633.1 | 5 - 6 | 508 | 108 | ||
| NM_016617.3 | NP_057701.1 | 5 - 6 | 336 | 108 | ||
| NR_104584.1 | 6 - 7 | 397 | 108 | |||
| GenBank mRNA | AB154404.1 | - | 4 - 5 | 187 | 108 | |
| AF208844.1 | - | 5 - 6 | 226 | 108 | ||
| AK095490.1 | - | 5 - 6 | 256 | 108 | ||
| AL515797.3 | - | 4 - 5 | 188 | 108 | ||
| BC005193.1 | - | 5 - 6 | 253 | 3829206 | 108 | |
| BI458294.1 | - | 5 - 6 | 307 | 108 | ||
| BP277186.1 | - | 5 - 6 | 336 | 108 | ||
| BU589105.1 | - | 6 - 7 | 313 | 108 | ||
| CR457189.1 | - | 4 - 5 | 187 | 108 | ||
| DA483023.1 | - | 5 - 6 | 256 | 108 | ||
More Information
Set Membership: |
|

UFM1
Genomic Map