Assay Details
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286822.1 | NP_001273751.1 | 2 - 3 | 460 | 104 | |
GenBank mRNA | AK298557.1 | - | 2 - 3 | 248 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus
positive regulation of protein ubiquitination
negative regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of cellular protein catabolic process
p53 binding
molecular_function
protein binding
protein import into nucleus
positive regulation of protein ubiquitination
negative regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of cellular protein catabolic process
p53 binding
molecular_function
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000474737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000480234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | ENST00000494690.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008390.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | TC2100008390.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | NR_104597.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000330938.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000397886.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000397887.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | ENST00000445724.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | NM_001286822.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | NM_004339.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100156878.hg.1 | NR_104597.1 | Refseq |
