Assay Details
| Gene Symbol: ELOVL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104558.1 | NP_001098028.1 | 5 - 6 | 534 | 78 | |
| NM_001297617.1 | NP_001284546.1 | 8 - 9 | 932 | 78 | ||
| NM_001297618.1 | NP_001284547.1 | 6 - 7 | 539 | 78 | ||
| NM_024930.2 | NP_079206.2 | 6 - 7 | 585 | 78 | ||
| XM_005248606.4 | XP_005248663.1 | 5 - 6 | 1149 | 78 | ||
| XM_005248607.3 | XP_005248664.1 | 6 - 7 | 862 | 78 | ||
| XM_006714695.3 | XP_006714758.1 | 7 - 8 | 641 | 78 | ||
| XM_011543651.2 | XP_011541953.1 | 7 - 8 | 831 | 78 | ||
| XM_017009885.1 | XP_016865374.1 | 6 - 7 | 522 | 78 | ||
| XM_017009886.1 | XP_016865375.1 | 8 - 9 | 761 | 78 | ||
| XM_017009887.1 | XP_016865376.1 | 8 - 9 | 952 | 78 | ||
| GenBank mRNA | AB181393.1 | - | 6 - 7 | 526 | 78 | |
| BC094792.1 | - | 8 - 9 | 873 | 78 | ||
| BC130310.1 | - | 5 - 6 | 482 | 40146341 | 78 | |
| BC130312.1 | - | 5 - 6 | 482 | 40146343 | 78 | |
More Information
Set Membership: |
|

ELOVL7
Genomic Map