| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
ENST00000396496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
ENST00000396499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
ENST00000511257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
NM_176816.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
ENST00000396496.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
ENST00000396499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
ENST00000511257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
NM_176816.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500176544.hg.1 |
TC0500010998.hg.1 |
Clariom D TC |